Learn More
Honeywell™ Perfect Fit™ Aramid Fiber Seamless Knit Gloves
Does not break down in high temperatures
Manufacturer:  Honeywell™ KV18AL100
 View more versions of this product
Catalog No. 19-050-597
Add to cart
Description
Description
Gloves do not provide thermal heat protection.
  • 100% Kevlar™ Aramid fibers brand seamless fiber construction provides exceptional cut, slash, and abrasion resistance
  • Construction provides excellent fit and durability
  • No seams to rub or split
  • Can be laundered to extend service life
Specifications
Specifications
Safety
automotive assembly, sheet Metal, Glass handling, Metal stamping, Metal fabrication, recycling, canning, injection moulding, Paper/pulp industry
Women's
Yellow
Level 3
Dupont™ Kevlar™ Aramid Fiber
Medium weight