Learn More
Honeywell™ Perfect Fit Aramid Fiber Sleeves
Honeywell offers a wide range of knitted arm protection in various sizes and lengths. DuPont™ Kevlar™ engineered fiber helps prevent cuts and slashes and is inherently flame-resistant. Aramid sleeves offer an excellent choice for the glass and steel industries.
Manufacturer:  Honeywell™ KVS210
 View more versions of this product
Catalog No. 19-049726
Add to cart
Description
Description
Perfect Fit - Kevlar™ Aramid fibers Sleeves feature DuPont™ Kevlar™ Aramid fibers brand fiber and are available in a variety of lengths, widths and configurations. Kevlar™ Aramid fibers engineered fiber is 5 times stronger than steel on an equal weight basis and provides excellent protection from cut, slash and abrasion hazards. Kevlar™ Aramid fibers is also flame resistant, and can withstand heat up to 900°F (400°C) without degrading.
Specifications
Specifications
Safety
10 in.
Dupont™ Kevlar™ aramid fibers
Sleeves, Cut resistant
Black
25.4cm
Metal sheet handling, recycling, automotive assembly, blade handling, knife sharpening, Glass handling
2-ply