Learn More
Honeywell™ Perfect Fit™ HPPE Cut Resistant White Gloves - 13 Gauge
Cut resistant synthetic fiber is lightweight, low-linting, and 10 times stronger than steel on an equal weight basis. PowerCuff™ system helps to prevent cuff blow-out after multiple washes.
$13.25
Specifications
ANSI Abrasion Rating ANSI 1
Color White
Cut Resistance Rating ANSI A4
Glove Cuff Style Knit
Material HPPE
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-049-895D
View Documents
Honeywell™
PF13L
Large
Each for $13.25
Add to cart
19-049-895E
View Documents
Honeywell™
PF13XL
X-Large This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 13-gauge HPPE synthetic fiber gloves are lightweight and low linting
  • Cut-resistant synthetic fiber is 10 times stronger than steel on an equal weight basis
  • PowerCuff™ system helps to prevent cuff blow-out after multiple washes
  • Machine washable at temperatures up to 120°F
Specifications
Specifications
ANSI 1
ANSI A4
HPPE
13
White
Knit
PF13