Learn More
Cadence Science™ Perfektum™ Rustless Steel Syringe Needles
Robust stainless-steel design offers accurate, reproducible injections
Manufacturer:  Cadence Science™ 4031/DISCBYVEND
 View more versions of this product
Catalog No. 22-023102
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
  • Chrome-plated brass female luer hub
  • Corrosion-resistant Type 304 stainless steel needle
  • Regular medical point
  • Variety of sizes and gauges available
  • Approved for laboratory use only
Specifications
Specifications
Stainless Steel
For laboratory use only
Hypodermic Needle
Regular Medical
Non-sterile
24 G
1.5 in.
38 mm
Female Luer