Perfex™ Floor Brushes

Manufacturer: Perfex Corp.Supplier Diversity Partner 2518R

 View more versions of this product

Catalog No. 19-042-016B

  Check Availability
Add to Cart

Required Accessories : Requires 19-042-037 or 19-042-040 adjustable polymer handle (sold separately).

    Specifications

    Brush Style Stiff-sweeping bristle
    Color Red
    For Use With (Equipment) Polymer adjustable handle
    Length (English) 18 in.
    Length (Metric) 46cm
    Material Polypropylene Fiber
    For Use With (Application) For sweeping heavy or coarse material and for removing difficult dirt and deposits from unfinished surfaces, Wood, concrete, blacktop
    Product Type Stiff Sweep Floor Brush

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit