Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech.

Click to view available options
Quantity:
10g
50g
374-76-5
C9F18
450.07
MFCD00066614
MGOFOLPXKULBGG-UHFFFAOYSA-N
perfluoro-1,3,5-trimethylcyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl cyclohexane,acmc-1ajc4,nonafluoro-1,3,5-tris-trifluoromethyl-cyclohexane,1,2,2,3,4,4,5,6,6-nonafluoro-1,3,5-tris trifluoromethyl cyclohexane,cyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl,1,1,2,3,3,4,5,5,6-nonakis fluoranyl-2,4,6-tris trifluoromethyl cyclohexane
2776392
1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
C1(C(C(C(C(C1(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F
374-76-5
C9F18
450.07
MFCD00066614
MGOFOLPXKULBGG-UHFFFAOYSA-N
perfluoro-1,3,5-trimethylcyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl cyclohexane,acmc-1ajc4,nonafluoro-1,3,5-tris-trifluoromethyl-cyclohexane,1,2,2,3,4,4,5,6,6-nonafluoro-1,3,5-tris trifluoromethyl cyclohexane,cyclohexane,1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris trifluoromethyl,1,1,2,3,3,4,5,5,6-nonakis fluoranyl-2,4,6-tris trifluoromethyl cyclohexane
2776392
1,1,2,3,3,4,5,5,6-nonafluoro-2,4,6-tris(trifluoromethyl)cyclohexane
C1(C(C(C(C(C1(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F)(F)F)(C(F)(F)F)F
Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point -68°C
Quantity 10g
Boiling Point 125°C to 128°C
Formula Weight 450.07
Grade Technical
Chemical Name or Material Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers