Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Perfluoro(1,3-dimethylcyclohexane), tech. 90%

Manufacturer:  Alfa Aesar™ 4547718

Quantity 50g

 View more versions of this product

Catalog No. AA4547718


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

335-27-3
400.06
2064147
Technical Grade
C8F16
MFCD00001469
Perfluoro (1,3-dimethylcyclohexane)

Specifications

1.828g/mL
Odorless
101° to 102°C
C8F16
400.06
2064147
90%
335-27-3
Liquid
−55°C
50g
MFCD00001469
Perfluoro (1,3-dimethylcyclohexane)
Technical Grade

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit