Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Perfluoro-1-iododecane, 97%

Click to view available options
Quantity:
1g
5g
25g
423-62-1
C10F21I
645.981
MFCD00001065
UDWBMXSQHOHKOI-UHFFFAOYSA-N
perfluorodecyl iodide,1-iodoperfluorodecane,perfluoro-1-iododecane,henicosafluoro-10-iododecane,heneicosafluorodecyl iodide,heneicosafluoro-1-iododecane,decane, heneicosafluoro-1-iodo,iodoperfluorodecane,henicosafluorodecyl iodide,1-iodohenicosafluorodecane
67920
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
423-62-1
C10F21I
645.981
MFCD00001065
UDWBMXSQHOHKOI-UHFFFAOYSA-N
perfluorodecyl iodide,1-iodoperfluorodecane,perfluoro-1-iododecane,henicosafluoro-10-iododecane,heneicosafluorodecyl iodide,heneicosafluoro-1-iododecane,decane, heneicosafluoro-1-iodo,iodoperfluorodecane,henicosafluorodecyl iodide,1-iodohenicosafluorodecane
67920
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-henicosafluoro-10-iododecane
C(C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F
Quantity 25g
Boiling Point 195°C to 200°C
Melting Point 63°C to 67°C
Beilstein 1810506
Formula Weight 645.98
Percent Purity 97%
Chemical Name or Material Perfluoro-1-iododecane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MD8710000

TSCA : TSCA