Learn More
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
Supplier:  Thermo Scientific™ TS65193
 View more versions of this product
Catalog No. PI65193
Add to Cart
Description
Description
Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.
  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GC/MS
10 x 1 mL
Corrosive
Pentafluoro-propanoic acid anhydride
Room Temperature
98 to 100%
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.