Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-Compound FC-43, ACROS Organics™

Click to view available options
Quantity:
10g
100g
250g
Packaging:
Glass bottle
311-89-7
C12F27N
671.096
MFCD00000436
RVZRBWKZFJCCIB-UHFFFAOYSA-N
perfluorotributylamine,tris perfluorobutyl amine,tri perfluorobutyl amine,tris nonafluorobutyl amine,heptacosafluorotributylamine,fluorinert,fluosol 43,perfluorotri-n-butylamine,fluorocarbon fc 43,tri nonafluorobutyl amine
9397
CHEBI:38854
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
311-89-7
C12F27N
671.096
MFCD00000436
RVZRBWKZFJCCIB-UHFFFAOYSA-N
perfluorotributylamine,tris perfluorobutyl amine,tri perfluorobutyl amine,tris nonafluorobutyl amine,heptacosafluorotributylamine,fluorinert,fluosol 43,perfluorotri-n-butylamine,fluorocarbon fc 43,tri nonafluorobutyl amine
9397
CHEBI:38854
1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Boiling Point 173.0°C to 181.0°C
Melting Point -52.0°C
Infrared Spectrum Authentic
Viscosity 2.3-3.3 mm2/s (25°C)
Color Colorless
Specific Gravity 1.88
Refractive Index 1.291
Quantity 100g
Packaging Glass bottle
Density 1.8800g/mL
Solubility Information Solubility in water: insoluble.
Formula Weight 670
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-43™
Show More Show Less

EINECSNumber : 206-223-1

RTECSNumber : YA1000000

TSCA : TSCA