Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-compound FC-72(TM), ACROS Organics™

Click to view available options
Quantity:
100g
250g
Packaging:
Glass bottle
355-42-0
C6F14
340
MFCD00000437
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
perfluorohexane,perflexane,tetradecafluorohexane,perfluoro-n-hexane,flutec pp1,hexane, tetradecafluoro,n-tetradecafluorohexane,fluorinert fc72,perfluorohexanes,perflexane usan:inn
9639
CHEBI:39427
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
355-42-0
C6F14
340
MFCD00000437
ZJIJAJXFLBMLCK-UHFFFAOYSA-N
perfluorohexane,perflexane,tetradecafluorohexane,perfluoro-n-hexane,flutec pp1,hexane, tetradecafluoro,n-tetradecafluorohexane,fluorinert fc72,perfluorohexanes,perflexane usan:inn
9639
CHEBI:39427
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
Specific Gravity 1.68
Melting Point -4.0°C
Boiling Point 51.0°C to 59.0°C
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
Density 1.6800g/mL
Color Colorless
Refractive Index 1.251
Viscosity 0.42 mm2/s (25°C)
Merck Index 15, 7271
Solubility Information Solubility in water: insoluble
Formula Weight 340
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-72&asterisk;
Show More Show Less

GHS H Statement:
May cause long lasting harmful effects to aquatic life.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-585-0

RTECSNumber : MO4310000

TSCA : TSCA