Alfa Aesar™ Perfluoro-tert-butanol, 99%

Manufacturer: Alfa Aesar™ B2244203

Quantity 1g

 View more versions of this product

Catalog No. AAB2244203

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2378-02-1
    Molecular Formula C4HF9O
    Linear Formula (CF3)3COH
    Formula Weight 236.04
    MDL Number MFCD00042092
    UN Number UN2810
    Beilstein 1841640
    Synonym 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol; Nonafluoro-tert-butyl alcohol
    Chemical Name or Material Perfluoro-tert-butanol

    Specifications

    Formula Weight 236.04
    Odor Characteristic
    MDL Number MFCD00042092
    UN Number UN2810
    Synonym 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol; Nonafluoro-tert-butyl alcohol
    Health Hazard 1 Danger
    Melting Point −17°C
    Health Hazard 3 GHS P Statement
    P260-P280f-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number UB6452700
    Quantity 1g
    Molecular Formula C4HF9O
    Linear Formula (CF3)3COH
    CAS 2378-02-1
    Beilstein 1841640
    Health Hazard 2 GHS H Statement
    H331-H315-H319-H335
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 44° to 45°C
    Chemical Name or Material Perfluoro-tert-butanol
    Assay 99%

    GHS H Statement
    H331-H315-H319-H335
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280f-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit