Learn More
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
$193.00
Specifications
For Use With FID and ECD
Standard Concentration 98 to 100%
View More Specs
Products 1
Catalog Number Mfr. No. Standard Components Price Quantity & Availability  
Catalog Number Mfr. No. Standard Components Price Quantity & Availability  
PI44211
View Documents
Thermo Scientific™
TS44211
1-(2,2,3,3,4,4,4-Heptafluoro-1-oxobutyl)-1 H-imidazole
Each for $193.00
Add to cart
Description
Description
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI). Compared to acid chlorides and anhydrides, the reactions of fluorinated acylimidazoles are smooth and quantitative, and produce no acid byproducts. The principal byproduct is imidazole, which is relatively inert. The volatile and stable fluoroacyl derivatives are ideal for FID and electron-capture detection (ECD) techniques.
Applications
Used in bifunctional derivatization schemes and in exchange reactions in which TMS derivatives are converted to HFB derivatives.
Hydroxyl groups of catecholamines are derivatized with TMSI, followed by conversion of the amines to acylamides with HFBI.
Tryptamine and metabolites present in spinal fluid have been analyzed by ECD using HFBI.
Specifications
Specifications
FID and ECD
98 to 100%
SDS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.