Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorodecalin, 90%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
306-94-5
C10F18
462.08
MFCD00010626
UWEYRJFJVCLAGH-UHFFFAOYSA-N
perfluorodecalin,perflunafene,octadecafluorodecahydronaphthalene,cis-perfluorodecalin,trans-perfluorodecalin,perfluorodecahydronaphthalene,naphthalene, octadecafluorodecahydro,f-dc,pp 5,perflunafenum latin
9386
CHEBI:38848
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F
306-94-5
C10F18
462.08
MFCD00010626
UWEYRJFJVCLAGH-UHFFFAOYSA-N
perfluorodecalin,perflunafene,octadecafluorodecahydronaphthalene,cis-perfluorodecalin,trans-perfluorodecalin,perfluorodecahydronaphthalene,naphthalene, octadecafluorodecahydro,f-dc,pp 5,perflunafenum latin
9386
CHEBI:38848
1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F
Specific Gravity 1.9
Refractive Index 1.3135 to 1.3155
Density 1.9000g/mL
Quantity 25g
Color Colorless
Infrared Spectrum Authentic
Melting Point -10.0°C
Flash Point 59°C
Boiling Point 142.0°C
Packaging Glass bottle
Assay Percent Range 90%
Merck Index 15,4215
Solubility Information Solubility in water: insoluble
Formula Weight 462.08
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Perfluorodecalin
Show More Show Less

EINECSNumber : 206-192-4

RTECSNumber : QJ3175000

TSCA : TSCA