Alfa Aesar™ Perfluorodimethylcyclobutane, mixture of 1,2 and 1,3 isomers, 97%

Manufacturer: Alfa Aesar™ 4303609

Quantity 10g

 View more versions of this product

Catalog No. AA4303609

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 28677-00-1
    Molecular Formula C6F12
    Linear Formula C4F6(CF3)2
    Formula Weight 300.05
    MDL Number MFCD00013737
    Chemical Name or Material Perfluorodimethylcyclobutane, Mixture of 1,2 and 1,3 isomers

    Specifications

    Formula Weight 300.05
    Refractive Index 1.2540
    Physical Form Liquid
    Boiling Point 45°C
    Molecular Formula C6F12
    Quantity 10g
    Density 1.620g/mL
    Linear Formula C4F6(CF3)2
    MDL Number MFCD00013737
    CAS 28677-00-1
    Chemical Name or Material Perfluorodimethylcyclobutane, Mixture of 1,2 and 1,3 isomers
    Assay 97%
  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit