Alfa Aesar™ Perfluoroheptanoic acid, 98+%

Manufacturer: Alfa Aesar™ A1209222

Quantity 100g

 View more versions of this product

Catalog No. AAA1209222

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 375-85-9
    Molecular Formula CF3(CF2)5CO2H
    Formula Weight 364.06
    MDL Number MFCD00039604
    UN Number UN3261
    Beilstein 1808210
    Synonym n-Tridecafluoroheptanoic acid
    Chemical Name or Material Perfluoroheptanoic acid

    Specifications

    Melting Point 31° to 36°C (lit)
    Formula Weight 364.06
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 100g
    CAS 375-85-9
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Boiling Point 89°C (30mmHg)
    Beilstein 1808210
    Synonym n-Tridecafluoroheptanoic acid
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula CF3(CF2)5CO2H
    MDL Number MFCD00039604
    UN Number UN3261
    Flash Point >110°C (230°F)
    Chemical Name or Material Perfluoroheptanoic acid
    Assay ≥98%

    Description

    Applications
    Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

    Solubility
    Insoluble in water.

    Notes
    Store at room temperature. Keep away from oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit