Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorohexyl bromide, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
335-56-8
C6BrF13
398.95
JTYRBFORUCBNHJ-UHFFFAOYSA-N
1-bromoperfluorohexane,perfluorohexyl bromide,perfluorohexylbromide,tridecafluorohexyl bromide,1-bromo-tridecafluorohexane,bromoperfluorohexane,acmc-1cmhf,1-bromotridecafluorohexane,tridecafluoro-1-bromohexane
92755
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
335-56-8
C6BrF13
398.95
JTYRBFORUCBNHJ-UHFFFAOYSA-N
1-bromoperfluorohexane,perfluorohexyl bromide,perfluorohexylbromide,tridecafluorohexyl bromide,1-bromo-tridecafluorohexane,bromoperfluorohexane,acmc-1cmhf,1-bromotridecafluorohexane,tridecafluoro-1-bromohexane
92755
1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F
Specific Gravity 1.871
Boiling Point 98.0°C to 100.0°C
Quantity 5g
Density 1.8710g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula F3C(CF2)4CF2Br
Formula Weight 398.95
Percent Purity 97%
Chemical Name or Material Perfluorohexyl bromide

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 206-391-6