Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perforin Mouse anti-Human, Porcine, Clone: dG9 (delta G9), eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14999480

 View more versions of this product

Catalog No. 50-139-77


Add to cart

Description

Description

Description: The dG9 antibody clone reacts with human perforin (pore-forming protein, pfp). Perforin is one of the cytolytic mediators present in the cytoplasmic granules of cytotoxic T lymphocytes (CTL) and natural killer cells (NK). Perforin is involved in the killing function by CTLs and NKs and has an important role in the immune response against tumors and virus infections. Applications Reported: This dG9 (delta G9) antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. (Fluorochrome conjugated dG9 (delta G9) is recommended for use in intracellular flow cytometry.). Applications Tested: This dG9 (delta G9) antibody has been tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

Perforin is one of the major cytolytic proteins of cytolytic granules. Perforin is a cytolytic mediator and is stored in and released by cytoplasmic granules. Moreover, perforin is involved in immune defense against tumors and virus infections as mediated by cytotoxic lymphocytes. Perforin is a 555 amino acid protein with a 21 amino acid signal peptide, and has a molecular weight of 70 to 75 kD. Perforin is a pore forming protein with a mechanism of transmembrane channel formation similar to C9, and homology between perforin and C9 have been demonstrated. Studies show that perforin is expressed only in killer cell lines and not in helper T lymphocytes or other tumor cells tested. Perforin is known to be a key effector molecule for T-cell- and natural killer-cell-mediated cytolysis. Defects in the perforin gene cause familial hemophagocytic lymphohistiocytosis type 2 (HPLH2), a rare and lethal autosomal recessive disorder of early childhood. Alternative splicing results in multiple transcript variants of perforin.
Specifications

Specifications

Perforin
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P14222
cytolysin; FLH2; HPLH2; lymphocyte pore forming protein; lymphocyte pore-forming protein; perforin 1 (pore forming protein); perforin-1; PFN1; PFP; RP11-710A11.3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Porcine
Immunohistochemistry, Immunoprecipitation
dG9 (delta G9)
Unconjugated
PRF1
Liquid
PRF1
IgG2b, kappa
25 μg
4° C
Primary
396595, 5551
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.