Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perforin Rat anti-Mouse, Janelia Fluor 646, Clone: CB5.4, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP197512JF646
Description
Perforin Monoclonal antibody specifically detects Perforin in Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Perforin | |
CB5.4 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Purified | |
PRF1 | |
Recombinant mouse perforin (aa 98-534). | |
Protein G purified | |
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
Primary | |
5551 | |
Mouse |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
Rat | |
IgG2a | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Recognizes mouse perforin (region aa 402-534). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title