Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Hidex™ Cut-Resistant Gloves

$114.10 - $116.00

Specifications

Color Black, Blue
Cut Resistance Rating Level 5
Material SuperFabric™, Synthetic Leather
For Use With (Application) Wood handling, automotive assembly, Glass handling, Metal fabrication, construction, industrial, general work
Glove Cuff Style Hook and loop
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-161-842D
View Documents
HexArmor™
4018XL
X-Large Pair for $114.10
Add to cart
 
19-161-842E
View Documents
HexArmor™
4018XXL
2X-Large Pair for $116.00
Add to cart
 
19-161-842A
View Documents
HexArmor™
4018S
Small Each for $114.10
Add to cart
 
Specifications

Specifications

Black, Blue
SuperFabric™, Synthetic Leather
Hook and loop
Level 5
Wood handling, automotive assembly, Glass handling, Metal fabrication, construction, industrial, general work
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit