Thermo Scientific™ Calibration Validation Carousels
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money.
Manufacturer: Thermo Scientific™ 840212900
 View more versions of this product
Catalog No. 14-380-440
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description NIST traceable
    For Use With Evolution 201, 220, 260 Bio and 300
    Includes Calibration Validation Carousel with Smart Base
    Item Description 840-212900
    Type Pharmacopia Peformance Validation - USP
    Certifications/Compliance All standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
    Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated confirguration for hands-free performance verification of your Thermo Scientific™ spectrophotometer.
    • Simplify qualification procedures by automating performance verification of UV-Visible spectrophotometers
    • Each CVC has a unique serial number matched automatically to the corresponding data files containing standard values and instrument specifications
    • Generated result reports are suitable for printing or archival purposes
    • Included standards are traceable to either NIST™ or NPL primary standards
    • CVCs are available in standard or pharmacopeia compliant configurations
    Comprehensive Performance Verification Testing
    • Wavelength Accuracy and Repeatability
    • Photometric Accuracy and Repeatability
    • Stray Light
    • Resolution
    • Photometric Noise
    • Photometric Stability or Drift
    • Baseline Flatness
    Save Time, Reduce Errors
    • Eliminates need to copy standard values from certificates
    • Eliminates need to analyze standards individually
    • Eliminates need to match standards to data
    • Eliminates need for manual calculations
    • Eliminates need for manual transcription and reporting of results
    Ordering Information:
    Unit is shipped in a plastic case for optimal protection during storage. Calibration Validation Carousels can be used with INSIGHT, INSIGHT Security, VISIONpro™, VISIONsecurity™ and applicable Local Control software. Pharmacopeia-compliant CVCs are compatible with the Evolution 600, 300, 260 Bio, 220 and 201 UV-Visible spectrophotometers. Standard configuration CVCs are additionally compatible with the AquaMate Plus, BioMate 6, Evolution 160, Helios Omega, Helios Zeta and UV-10 UV-Visible spectrophotometers.
  • Description & Specifications
Related Products
Product Content Feedback