Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perilipin-1 Mouse anti-Human,Bovine, KHL, Clone: Peri 112.17.1, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Perilipin-1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19421MI
View Documents
MilliporeSigma
MABS1942100UG
100 μg
Each for $372.00
Add to cart
 
MABS19422MI
View Documents
MilliporeSigma
MABS194225UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Perilipin-1 Monoclonal antibody specifically detects Perilipin-1 in Human,Bovine samples. It is validated for Electron Microscopy, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Perilipin-1
KHL
Mouse
Lipid droplet-associated protein
KLH-conjugated linear peptide corresponding to first 20 amino acids from the N-terminal region of human Perilipin-1.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Purified
Human, Bovine
PLIN1, PERI, PLIN
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit