Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perilipin Rabbit anti-Human, Clone: JM10-96, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP267037
Description
Perilipin Monoclonal antibody specifically detects Perilipin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Perilipin | |
JM10-96 | |
Unconjugated | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein | |
Rabbit | |
IgG | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human, Mouse |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
Purified | |
PLIN1 | |
Full length protein | |
Protein A purified | |
Diabetes Research, Lipid and Metabolism, Lipid Droplets | |
Primary | |
5346 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title