Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic acid, Acros Organics

Click to view available options
Quantity:
25g
100g
500g
10450-60-9
H5IO6
227.938
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid,periodic acid,tetrahydroxyiodous acid,acid, periodic,paraperiodic acid,pentahydroxy-l5-iodane oxide,unii-ak1d44l87g,pentahydroxidooxidoiodine,hexaoxoiodic 5-acid,periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O
10450-60-9
H5IO6
227.938
TWLXDPFBEPBAQB-UHFFFAOYSA-N
orthoperiodic acid,periodic acid,tetrahydroxyiodous acid,acid, periodic,paraperiodic acid,pentahydroxy-l5-iodane oxide,unii-ak1d44l87g,pentahydroxidooxidoiodine,hexaoxoiodic 5-acid,periodic acid, cryst
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O
Quantity 500g
Linear Formula H5IO6
Formula Weight 227.94
Grade Analysis
Solubility Solubility in water: 3 kg/l (20°C). Other solubilities: soluble in alcohol, slightly soluble in ether
Melting Point 122.0°C
Chemical Name or Material Periodic Acid