Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Periodic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
100g
Packaging:
Wide Mouth Amber Glass Bottle
10450-60-9
H5IO6
227.938
TWLXDPFBEPBAQB-UHFFFAOYSA-N
para-Periodic Acid
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O
10450-60-9
H5IO6
227.938
TWLXDPFBEPBAQB-UHFFFAOYSA-N
para-Periodic Acid
25289
CHEBI:29150
pentahydroxy(oxo)-$l^{7}-iodane
OI(=O)(O)(O)(O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 25g
Formula Weight 227.96
Packaging Wide Mouth Amber Glass Bottle
Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Periodic Acid