Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peripherin Mouse anti-Human, Mouse, Rat, HRP, Clone: OTI1E10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273337H
Description
Peripherin Monoclonal specifically detects Peripherin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Peripherin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PRPH | |
Full length recombinant protein of human Peripherin (NP_006253) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E10 | |
Western Blot, Immunohistochemistry | |
Neurofilament 4, neurofilament 4 (57kD), peripherin, PRPH1NEF4 | |
Mouse | |
Affinity Purified | |
Cancer, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
5630.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title