Learn More
Peripherin Rabbit anti-Human,Mouse,Rat, KHL, Clone: 1D4, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB117425UL
 View more versions of this product
Catalog No. ZRB117425MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Peripherin Monoclonal antibody specifically detects Peripherin in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Peripherin
1D4
KHL
Purified
PRPH, NEF4, PRPH1
KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Peripherin.
Target molecular weight: 53.65 kDa calculated; ∼53 kDa observed.
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_006253.2
Neurofilament 4
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
Human, Mouse, Rat