Learn More
Brady™ PermaShield™ Thermal Transfer Printable Labels
For general purpose identification including Asset tracking and Rating plates. Brady™ PermaShield™ Thermal Transfer Printable Labels are made of metallized Polyester with overlaminate.
$491.14 - $696.27
Specifications
Product Type Thermal Transfer Printer Label
Format Roll
Material Metallized Polyester (B-428), Over-laminate (B-966B)
Color Silver
Thickness (English) 0.0073 in.
View More Specs
Products 3
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Price Quantity & Availability  
19-109-303
View Documents
Brady™
THT224282
1.25 x 0.75 in. (W x H)
Each for $491.14
Add to cart
19-109-305
View Documents
Brady™
THT244281
2.75 x 1.75 in. (W x H)
Each for $696.27
Add to cart
19-109-304
View Documents
Brady™
THT234282
2 x 1 in. (W x H)
Each for $663.08
Add to cart
Description
Description
  • Made of metallized polyester with overlaminate so they remain clear and easy to read even in harsh environments
  • Material is UL Recognized, CSA Approved, AGA Approved, Halogen-Free (DIN VDE 0472/parts 815)
Specifications
Specifications
Thermal Transfer Printer Label
Metallized Polyester (B-428), Over-laminate (B-966B)
0.0073 in.
Asset Tracking, Rating Plates, General Purpose Identification
BBP™81 Label Printer, IP Series, BRADYPRINTER™ PR Plus Series Printer
Roll
Silver
Rectangle
Gloss