Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ PermaSleeve™ Heat-Shrink Polyolefin BMP51/BMP53/BMP41 Label Maker Cartridge

For use in harsh environments. Brady™ PermaSleeve™ Heat-Shrink Polyolefin BMP51/BMP53/BMP41 Label Maker Cartridges are durable and the shrink rate saves time during installation.

$66.66 - $141.37

Specifications

Product Type Label Maker Cartridge
Material Polyolefin (B-342)
Color Black, White
For Use With (Application) Wire and cable marking
Description Shrink Ratio: 3:1
View More Specs

Products 4
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19-109-475
View Documents
Brady™
M187075342
0.335 in.
Each for $66.66
Add to cart
 
19-109-474
View Documents
Brady™
M125075342
0.235 in.
Each for $74.47
Add to cart
 
19-109-476
View Documents
Brady™
M250075342
0.439 in.
Each for $98.22
Add to cart
 
19-109-477
View Documents
Brady™
M375075342
0.645 in.
Each for $141.37
Add to cart
 
Description

Description

  • 3:1 heat shrinkable sleeve fits snugly around wire
  • Chemical, corrosion and UV resistant
  • One cartridge contains 80 labels
Specifications

Specifications

Label Maker Cartridge
Black, White
Shrink Ratio: 3:1
BMP™51, BMP™53, BMP™41 Printers
Polyolefin (B-342)
Wire and cable marking
Matte
0.75 ft.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit