Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxidase from Horseradish, TCI America™

Click to view available options
Quantity:
100 mg
1 g
9003-99-0
9003-99-0
Quantity 100 mg
Physical Form Crystalline Powder

Recommended Storage : <0°C