Learn More
Aat BioquestSupplier Diversity Partner Amplite™ Fluorimetric Peroxidase (HRP) Assay Kit *Red Fluorescence*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11552
Peroxidase is a small molecule (MW ~40 KD) that can usually be conjugated to an antibody in a 4:1 ratio. Due to its small size, it rarely causes steric hindrance problem with antibody/antigen complex formation.
Catalog No. 50-195-3363