Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Peroxiredoxin 3
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094858
View Documents
Novus Biologicals
NBP303779100UL
100 μg
Each for $320.71
Add to cart
 
NB094859
View Documents
Novus Biologicals
NBP30377920UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Peroxiredoxin 3 Polyclonal antibody specifically detects Peroxiredoxin 3 in Human, Mouse samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

Peroxiredoxin 3
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
10935
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Human, Mouse
Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial
Recombinant fusion protein containing a sequence corresponding to amino acids 63-256 of human Peroxiredoxin 3 (NP_006784.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit