Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peroxiredoxin 4 Mouse anti-Human, Clone: CPTC-PRDX4-2, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Peroxiredoxin 4
Clone CPTC-PRDX4-2
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122459
View Documents
Novus Biologicals
NBP30749310UG
100 μg
Each for $352.89
Add to cart
 
NB122460
View Documents
Novus Biologicals
NBP30749320UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Peroxiredoxin 4 Monoclonal antibody specifically detects Peroxiredoxin 4 in Human samples. It is validated for ELISA
Specifications

Specifications

Peroxiredoxin 4
0.2 mg/ml
ELISA
Purified
RUO
Human
Antioxidant enzyme AOE372, AOE37-2PRX-4, EC 1.11.1.15, peroxiredoxin 4, Peroxiredoxin IV, peroxiredoxin-4, prx4, prx-4, Prx-IV, thioredoxin peroxidase (antioxidant enzyme), Thioredoxin peroxidase AO372, Thioredoxin-dependent peroxide reductase A0372
Recombinant full-length human Peroxiredoxin 4 protein
Monoclonal
Protein A or G purified
CPTC-PRDX4-2
ELISA
Unconjugated
Mouse
Signal Transduction
10 mM PBS with 0.05% BSA
10549
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit