Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 6 Antibody (22E7), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259436AF488
This item is not returnable.
View return policy
Description
Peroxiredoxin 6 Monoclonal antibody specifically detects Peroxiredoxin 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Peroxiredoxin 6 | |
Monoclonal | |
Alexa Fluor 488 | |
1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX | |
Recombinant human Peroxiredoxin 6 (1-224aa) purified from E. coli | |
0.1 mL | |
Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Immunology, Signal Transduction | |
9588 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
220000000 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction