Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin 6 Rabbit anti-Human, Mouse, Rat, Clone: 1A9W5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Peroxiredoxin 6 |
---|---|
Clone | 1A9W5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Peroxiredoxin 6 Monoclonal antibody specifically detects Peroxiredoxin 6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Peroxiredoxin 6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Immunology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
9588 | |
IgG | |
Affinity purified |
1A9W5 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRX | |
A synthetic peptide corresponding to a sequence within amino acids 125-224 of human Peroxiredoxin 6 (P30041). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title