Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peroxiredoxin, bacteria-like protein Mouse anti-Parasite, DyLight 405, Clone: V71, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259672V
Description
Peroxiredoxin, bacteria-like protein Monoclonal specifically detects Peroxiredoxin, bacteria-like protein in Parasite samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Peroxiredoxin, bacteria-like protein | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Peptide Sequence specific to peroxiredoxin antigen from Tetracapsuloides bryosalmonae (fish parasite). | |
0.1 mL | |
Virology Bacteria and Parasites | |
9682623 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
V71 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Parasite | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)