Learn More
Aviva Systems Biology Corp PERP Antibody - middle region (ARP49688_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49688_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PERP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PERP. Alternate names: KCP1, KRTCAP1, PIGPC1, RP3-496H19.1, THW, dJ496H19.1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60144