Learn More
Persephin Rat anti-Mouse, Alexa Fluor 700, Clone: RM0306-6H29, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211851AF700
Catalog No. NB211851700
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Persephin Monoclonal antibody specifically detects Persephin in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Persephin
RM0306-6H29
Western Blot
Purified
PSPN
Mouse Persephin
Protein G purified
Biologically Active Proteins
Primary
5623
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
persephin, PSP
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
Mouse