Learn More
Brady™ Personal Basic Lockout Kit with Keyed-Alike Steel Padlocks
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Personal Basic Lockout Kit with Keyed-Alike Steel Padlocks is a basic kit that includes all of the devices needed to lockout common breakers, valves and plugs.
Manufacturer:  Brady™ 104796
Catalog No. 19-104-151
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical & mechanical risk lockouts
7.1 in.
5.5 in.
14 in.
Safety
Black/red
(1) lockout box; (1) no-hole circuit breaker, 277v; (1) multi-pole circuit breaker; (1) gate valve lockout, 1-2.5 in.; (1) small ball valve lockout; (1) plug lockout, 110v; (2) 1 in. lockout hasps; (2) lockout tags; (2) kd steel padlock, 0.75 in. shackle, red
Polypropylene with nickle-plated clasps
5.5 in. x 14 in. x 7.1 in.