Learn More
Brady™ Personal Breaker Lockout Kit w/ 3 Keyed-Alike Safety Padlocks
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Personal Breaker Lockout Kit is a great solution that covers 120/277V and 480/600V breakers in single- and multi-pole configurations. A successful program begins with the right devices in the right place.
Manufacturer:  Brady™ 105964
Catalog No. 19-104-162
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
7.1 in.
5.5 in.
14 in.
Safety
Red/white on black
(1) lockout box; (2) no-hole circuit breaker, 277v; (2) no-hole circuit breaker, 480/600v; (2) multi-pole circuit breaker; (3) lockout hasps; (3) lockout tags; (3) ka safety padlocks, 1.5 in. shackle, red
Polypropylene
5.5 in. x 14 in. x 7.1 in.