Learn More
Brady™ Personal Breaker Lockout Pouch w/ 2 Keyed-Alike Steel Padlocks
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™Personal Breaker Lockout Pouch is a great solution that covers 120/277V and 480/600V breakers in single- and multi-pole configurations. A successful program begins with the right devices in the right place.
Manufacturer:  Brady™ 105968
Catalog No. 19-104-166
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical risk lockouts
4.75 in.
2 in.
7.5 in.
Safety
Black on red
(1) lockout bag; (1) no-hole circuit breaker, 277v; (1) no-hole circuit breaker, 480/600v; (1) multi-pole circuit breaker; (2) lockout hasps; (2) lockout tags; (2) ka steel padlocks
Nylon
2 in. x 7.5 in. x 4.75 in.