Learn More
Brady™ Personal Valve and Electrical Kit w/ 3 Keyed-Alike Steel Padlocks
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Personal Valve and Electrical Kit is part of a safety plan to protect workers and the worksite using lockout products. Lockout devices are an important part of safety protocol.
Manufacturer:  Brady™ 105956
Catalog No. 19-104-154
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
1/Kit
Electrical & mechanical risk lockouts
7.8 in.
7.3 in.
16.2 in.
Safety
Black/red
(1) lockout box; (1) adjustable gate valve lockout, 1-6.5 in.; (1) small ball valve lockout; (1) 3-in-1 plug lockout; (1) all purpose cable lockout; (2) no-hole circuit breaker, 277v; (1) no-hole circuit breaker, 480/600v; (2) multi-pole circuit breaker; (3) lockout hasps; (3) lockout tags; (3) ka steel padlocks; (1) base cover plug valve lockout, 7/8 in.; (1) base cover plug valve lockout, 1-3/8 in.
Polypropylene
7.3 in. x 16.2 in. x 7.8 in.