Learn More
AccuTec Blades Personna™ Single Edge Blades
AccuTec Blades Personna™ Single Edge Blades provide a product solution for any need
Manufacturer:  AccuTec Blades 620317
 View more versions of this product
Catalog No. 17-989-128
Add to cart
Description
Description
  • Used in medical device manufacturing and various applications
  • High Carbon Steel blades
  • Aluminum or steel back
  • Available unwrapped, or wrapped in protective paper
Specifications
Specifications
2-Facet
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
1.50 in.
0.30 mm
Non-sterile
Carbon Steel
Aluminum
38.10 mm
5625/Cs.