Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cypermethrin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

$55.20 - $126.30

Specifications

Product Line PESTANAL
Grade Analytical Standard
Linear Formula C22H19 Cl2 NO3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-102-0033
View Documents
MilliporeSigma Supelco
458352ML
2 mL
Each for $55.20
Add to cart
 
11-101-8904
View Documents
MilliporeSigma Supelco
4583510ML
10 mL
Each for $126.30
Add to cart
 
Description

Description

Agriculture and Environmental
Specifications

Specifications

PESTANAL
Analytical Standard
C22H19 Cl2 NO3
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

SDS
Product Certifications

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit