Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dichlon, MilliporeSigma™ Supelco™

Dichlon is an agricultural fungicide of the quinone class, widely used against a variety of plant pathogenic fungi.

Manufacturer:  MilliporeSigma Supelco 45434250MG

 View more versions of this product

Catalog No. 11-102-0579


Add to cart

Description

Description

Agriculture
Specifications

Specifications

117-80-6
MFCD00001677
227.04
Analytical Standard
250 mg
Analytical Standard
C10H4 Cl2O2
2,3-Dichloro-1,4-naphthoquinone; Dichlon
Neat
194°C to 197°C
PESTANAL
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : QL7525000

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH