Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dichlon, MilliporeSigma™ Supelco™

Dichlon is an agricultural fungicide of the quinone class, widely used against a variety of plant pathogenic fungi.

$24.65 - $26.40

Specifications

Product Line PESTANAL
Grade Analytical Standard
Linear Formula C10H4 Cl2O2
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-102-0605
View Documents
MilliporeSigma Supelco
454341EA
1 Each
Each for $24.65
Add to cart
 
11-102-0579
View Documents
MilliporeSigma Supelco
45434250MG
250 mg
Each for $26.40
Add to cart
 
Description

Description

Agriculture
Specifications

Specifications

PESTANAL
Analytical Standard
C10H4 Cl2O2
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : QL7525000

SDS
Product Certifications

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit