Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Phenol, MilliporeSigma™ Supelco™

Specifications

Product Line PESTANAL
Density 1.071 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula C6H5 OH
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-9779
View Documents
MilliporeSigma Supelco
35952500MG
500 mg
Each for $63.90
N/A
Due to product restrictions, please to purchase or view availability for this product.
11-101-9043
View Documents
MilliporeSigma Supelco
359521G
1 g
Each for $110.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
Specifications

Specifications

PESTANAL
Analytical Standard
Neat
1.071 g/mL (at 25°C)
C6H5 OH
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : SJ3325000

SDS
Product Certifications

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit