Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Phthalimide, MilliporeSigma™ Supelco™

Manufacturer:  MilliporeSigma Supelco 36734100MG

Catalog No. 11-100-1860


Add to cart

Description

Description

Agriculture
Specifications

Specifications

85-41-6
MFCD00005881
147.13
Analytical Standard
100 mg
PESTANAL
C8H5 NO2
1,3-Dihydro-1,3-dioxoisoindole
Neat
232°C to 235°C
150°C
Analytical Standard
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : TI3920000

SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH