Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PET112 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549375

Catalog No. PA549375

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu- tRNA(Gln).
Specifications

Specifications

PET112
Polyclonal
PBS with 0.09% sodium azide
O75879
Glutamyl-tRNA(Gln) amidotransferase subunit B, mitochondrial, Glu-AdT subunit B, 635-, Cytochrome c oxidase assembly factor PET112 homolog, GATB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5188
Western Blot
Unconjugated
GATB
Liquid
GATB
KLH conjugated synthetic peptide between 25-60 amino acids from the N-terminal region of human PET112.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit