Learn More
Foxx Life Sciences EZBio 60mL PETG Media Bottles, Non-Sterile, Square Storage Bottles with 24-415mm Neck, Without Caps, 24/PK DFS Item
Manufacturer:  Foxx Life Sciences 1371113FLS
EZBio PETG Non-Sterile Media Bottles are perfect for storage and shipment reagents and media. Bottles withstand temperatures down to -40C for freezing applications. Shatter-resistant and thick-walled. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps, including options with vented membranes or molded hose barbs, are available separately.
Catalog No. 50-161-3223